Proteintech
Manufacturer Code:125061AP150UL
Catalog # 50-172-8031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLEK Fusion Protein Ag3187 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p47; P47 Platelet 47 kDa protein pleckstrin PLEK; Platelet 47 kDa protein; Pleckstrin
Gene Aliases: 2010300B13Rik; P47; PLEK
UniProt ID: (Human) P08567, (Mouse) Q9JHK5, (Rat) Q4KM33
Entrez Gene ID: (Human) 5341, (Mouse) 56193, (Rat) 364206
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.