Invitrogen
Manufacturer Code:PA571510
Catalog # PIPA571510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 125-154 amino acids from the Central region of human PLD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline phosphatase 6; Mitochondrial cardiolipin hydrolase; Mitochondrial cardiolipin hydrolase 31-- Choline phosphatase 6 Mitochondrial phospholipase MitoPLD Phosphatidylcholine-hydrolyzing phospholipase D6 Phospholipase D6 PLD 6 Protein zucchini homolog PLD6; Mitochondrial phospholipase; MitoPLD; mZuc; novel Phospholipase D Active site motif-containing protein; Phosphatidylcholine-hydrolyzing phospholipase D6; phospholipase D family, member 6; Phospholipase D6; PLD 6; Protein zucchini homolog
Gene Aliases: 4933433K01Rik; Gm10; mitoPLD; mZuc; PLD6; ZUC
UniProt ID: (Human) Q8N2A8, (Mouse) Q5SWZ9
Entrez Gene ID: (Human) 201164, (Mouse) 194908
Molecular Function:
RNA binding protein
endoribonuclease
hydrolase
lipase
nuclease
nucleic acid binding
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.