Invitrogen
Manufacturer Code:MA521431
Catalog # PIMA521431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C7 |
Immunogen | PLCL2 (AAH36392 121 a.a. &bsim 210 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Inactive phospholipase C-like protein 2; phospholipase C, epsilon 2; Phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLC-epsilon-2; PLC-L(2); PLC-L2; PLCE2 PLC-L(2) PLC-L2 PLC-epsilon-2 inactive phospholipase C-like protein 2 phospholipase C epsilon 2 phospholipase C-L2 phospholipase C-epsilon-2
Gene Aliases: KIAA1092; PLCE2; PLCL2
UniProt ID: (Human) Q9UPR0
Entrez Gene ID: (Human) 23228
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.