Invitrogen
Manufacturer Code:PA5110180
Catalog # PIPA5110180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 130-300 of human PLCL2 (NP_055999.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inactive phospholipase C-like protein 2; Inactive phospholipase C-like protein 2 KIAA1092 mKIAA1092 phospholipase C Phospholipase C-epsilon-2 Plce2 PLC-epsilon-2 PLC-L(2) PLCL2 PLC-L2 PRIP-2; phospholipase C, epsilon 2; Phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLC-epsilon-2; PLC-L(2); PLC-L2
Gene Aliases: KIAA1092; mKIAA1092; PLC-L2; PLCE2; PLCL2; PRIP-2
UniProt ID: (Human) Q9UPR0, (Mouse) Q8K394
Entrez Gene ID: (Human) 23228, (Mouse) 224860
Molecular Function:
G-protein modulator
calcium-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
hydrolase
lipase
phospholipase
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.