Novus Biologicals
Manufacturer Code:NBP213768
Catalog # NBP213768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: DMLESSQDNMRTSWVSQMFSEIDVDNLGHITLCNAVQCIRNLNPGLKTSK IELKFKELHKSKDKAGTEVTKEEFIE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.4.11 FLJ13484 inactive phospholipase C-like protein 2 KIAA1092PLCE2 phospholipase C epsilon 2 Phospholipase C-epsilon-2 Phospholipase C-L2 phospholipase C-like 2 PLC-epsilon-2 PLC-L(2) PLC-L2; Inactive phospholipase C-like protein 2; phospholipase C, epsilon 2; Phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLC-epsilon-2; PLC-L(2); PLC-L2
Gene Aliases: KIAA1092; PLCE2; PLCL2
UniProt ID: (Human) Q9UPR0
Entrez Gene ID: (Human) 23228
Molecular Function:
G-protein modulator
calcium-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
hydrolase
lipase
phospholipase
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.