Proteintech
Manufacturer Code:191431AP150UL
Catalog # 50-173-2356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLCH1 Fusion Protein Ag13607 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase eta-1; KIAA1069 Phospholipase C eta 1 Phospholipase C like protein 3 phospholipase C eta 1 PLC eta 1 PLC L3 PLCeta1 PLCH1 PLCL3; Phosphoinositide phospholipase C-eta-1; phospholipase C, eta 1; Phospholipase C-eta-1; phospholipase C-eta1a; phospholipase C-eta1b; phospholipase C-like 3; Phospholipase C-like protein 3; PLC eta 1; PLC-eta-1; PLC-L3
Gene Aliases: BC042549; KIAA1069; PLCeta1; PLCH1; PLCL3; RGD1561991
UniProt ID: (Human) Q4KWH8, (Mouse) Q4KWH5
Entrez Gene ID: (Human) 23007, (Mouse) 269437, (Rat) 310463
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.