Invitrogen
Manufacturer Code:PA553333
Catalog # PIPA553333
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PLCE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase epsilon-1; Pancreas-enriched phospholipase C; phosphoinositide phospholipase C; Phosphoinositide phospholipase C-epsilon-1; phosphoinositide-specific phospholipase C epsilon-1; phospholipase C, epsilon 1; Phospholipase C-epsilon-1; PLC-epsilon-1
Gene Aliases: KIAA1516; NPHS3; PLCE; PLCE1; PPLC
UniProt ID: (Human) Q9P212
Entrez Gene ID: (Human) 51196
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.