Invitrogen
Manufacturer Code:PA558264
Catalog # PIPA558264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PLCD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-4; hPLCD4; Phosphoinositide phospholipase C-delta-4; phospholipase C, delta 4; Phospholipase C-delta-4; PLC delta4; PLC-delta-4
Gene Aliases: PLCD4
UniProt ID: (Human) Q9BRC7
Entrez Gene ID: (Human) 84812
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.