Invitrogen
Manufacturer Code:PA568048
Catalog # PIPA568048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human PLCD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1; NDNC3 PLC-III 1-phosphatidylinositol-45-bisphosphate phosphodiesterase delta-1 PLC-delta-1 phosphoinositide phospholipase C-delta-1 phospholipase C-III; phosphoinositide phospholipase C; Phosphoinositide phospholipase C-delta-1; phospholipase C delta 1 Lf; phospholipase C delta 1 long form; phospholipase C, delta 1; Phospholipase C-delta-1; Phospholipase C-delta1; Phospholipase C-III; PLC-delta 1; PLC-delta-1; PLC-III
Gene Aliases: AW212592; C79986; NDNC3; PLC-III; Plc1; Plcd; PLCD1
UniProt ID: (Human) P51178, (Mouse) Q8R3B1, (Rat) P10688
Entrez Gene ID: (Human) 5333, (Mouse) 18799, (Rat) 24655
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.