Invitrogen
Manufacturer Code:PA5115769
Catalog # PIPA5115769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PLCD1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-1; 1-phosphatidylinositol 45-bisphosphate phosphodiesterase delta-1 1-phosphatidylinositol-45-bisphosphate phosphodiesterase delta-1 AW212592 C79986 NDNC3 phosphoinositide phospholipase C Phosphoinositide phospholipase C-delta-1 phospholipase C delta 1 phospholipase C delta 1 Lf phospholipase C delta 1 long form phospholipase C delta 1 Phospholipase C-delta1 phospholipase C-delta-1 phospholipase C-III Plc1 Plcd Plcd1 PLC-delta 1 PLC-delta-1 PLC-III; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1; phosphoinositide phospholipase C; Phosphoinositide phospholipase C-delta-1; phospholipase C delta 1 Lf; phospholipase C delta 1 long form; phospholipase C, delta 1; Phospholipase C-delta-1; Phospholipase C-delta1; Phospholipase C-III; PLC-delta 1; PLC-delta-1; PLC-III
Gene Aliases: AW212592; C79986; NDNC3; PLC-III; Plc1; Plcd; PLCD1
UniProt ID: (Human) P51178, (Mouse) Q8R3B1, (Rat) P10688
Entrez Gene ID: (Human) 5333, (Mouse) 18799, (Rat) 24655
Molecular Function:
G-protein modulator
calcium-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
hydrolase
lipase
phospholipase
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.