Invitrogen
Manufacturer Code:PA578109
Catalog # PIPA578109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human phospholipase C beta 3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3; Phosphoinositide phospholipase C-beta-3; phospholipase C beta 3; phospholipase C, beta 3 (phosphatidylinositol-specific); Phospholipase C-beta-3; PLC beta 3; PLC-beta-3; PLCB3 phospholipase C beta 3 (phosphatidylinositol-specific)
Gene Aliases: mKIAA4098; PLCB3
UniProt ID: (Human) Q01970, (Mouse) P51432
Entrez Gene ID: (Human) 5331, (Rat) 29322, (Mouse) 18797
Molecular Function:
G-protein modulator
calcium-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
hydrolase
lipase
phospholipase
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.