Invitrogen
Manufacturer Code:PA5103068
Catalog # PIPA5103068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PLCB3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-3; 1-phosphatidylinositol 45-bisphosphate phosphodiesterase beta-3 1-phosphatidylinositol-45-bisphosphate phosphodiesterase beta-3 EC 3.1.4.11 mKIAA4098 Phosphoinositide phospholipase C-beta-3 phosphoinositide-specific phospholipase c phospholipase C beta 3 phospholipase C beta 3 phospholipase C beta 3 (phosphatidylinositol-specific) phospholipase C-beta-3 PLC beta 3 PLCB3 PLCbeta3 PLC-beta3 PLC-beta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3; Phosphoinositide phospholipase C-beta-3; phospholipase C beta 3; phospholipase C, beta 3 (phosphatidylinositol-specific); Phospholipase C-beta-3; PLC beta 3; PLC-beta-3
Gene Aliases: mKIAA4098; PLCB3
UniProt ID: (Human) Q01970, (Mouse) P51432
Entrez Gene ID: (Human) 5331, (Rat) 29322, (Mouse) 18797
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.