Invitrogen
Manufacturer Code:PA548052
Catalog # PIPA548052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human PLC-beta 1 Lys27-Met245 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1; EIEE12 PI-PLC PLC-154 PLC-I PLC154 PLCB1A PLCB1B 1-phosphatidyl-D-myo-inositol-45-bisphosphate 1-phosphatidylinositol 45-bisphosphate phosphodiesterase beta-1 1-phosphatidylinositol-45-bisphosphate phosphodiesterase beta 1 1-phosphatidylinositol-45-bisphosphate phosphodiesterase beta-1 PLC-beta-1 in; inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; phosphoinositidase C; phosphoinositide phospholipase C; Phosphoinositide phospholipase C-beta-1; phospholipase C, beta 1 (phosphoinositide-specific); Phospholipase C-beta-1; phospholipase C-beta-1a; Phospholipase C-I; PLC-154; PLC-beta-1; PLC-I; triphosphoinositide phosphodiesterase
Gene Aliases: 3110043I21Rik; AI132408; EIEE12; KIAA0581; mKIAA0581; PI-PLC; PLC-154; PLC-beta-1; PLC-I; PLC154; Plcb; PLCB1; PLCB1A; PLCB1B
UniProt ID: (Human) Q9NQ66, (Mouse) Q9Z1B3
Entrez Gene ID: (Human) 23236, (Mouse) 18795
Molecular Function:
G-protein modulator
calcium-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
hydrolase
lipase
phospholipase
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.