Invitrogen
Manufacturer Code:PA526381
Catalog # PIPA526381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1148-1176 amino acids from the C-terminal region of human PLCB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1; EIEE12 PI-PLC PLC-154 PLC-I PLC154 PLCB1A PLCB1B 1-phosphatidyl-D-myo-inositol-45-bisphosphate 1-phosphatidylinositol-45-bisphosphate phosphodiesterase beta 1 1-phosphatidylinositol-45-bisphosphate phosphodiesterase beta-1 PLC-beta-1 inositoltrisphosphohydrolase monophosphatidylinositol phosphodieste; inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; phosphoinositidase C; Phosphoinositide phospholipase C-beta-1; phospholipase C, beta 1 (phosphoinositide-specific); Phospholipase C-beta-1; Phospholipase C-I; PLC-154; PLC-beta-1; PLC-I; triphosphoinositide phosphodiesterase
Gene Aliases: EIEE12; KIAA0581; PI-PLC; PLC-154; PLC-beta-1; PLC-I; PLC154; PLCB1; PLCB1A; PLCB1B
UniProt ID: (Human) Q9NQ66
Entrez Gene ID: (Human) 23236
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.