Novus Biologicals
Manufacturer Code:NBP29334720UL
Catalog # NB028579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1050-1150 of human PLCB3 (NP_000923.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-3; 1-phosphatidylinositol-45-bisphosphate phosphodiesterase beta-3 EC 3.1.4.11 FLJ37084 Phosphoinositide phospholipase C-beta-3 phospholipase C beta 3 (phosphatidylinositol-specific) Phospholipase C-beta-3 PLC beta 3 PLC-beta-3; Phosphoinositide phospholipase C-beta-3; phospholipase C, beta 3 (phosphatidylinositol-specific); Phospholipase C-beta-3; PLC beta 3; PLC-beta-3
Gene Aliases: PLCB3
UniProt ID: (Human) Q01970
Entrez Gene ID: (Human) 5331
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator guanyl-nucleotide exchange factor hydrolase lipase phospholipase signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.