Invitrogen
Manufacturer Code:PA568857
Catalog # PIPA568857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse PLBD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAMA-like protein 1; Lamina ancestor homolog 1; Phospholipase B domain-containing protein 1; Phospholipase B-like 1; Phospholipase B-like 1 chain A; Phospholipase B-like 1 chain B; Phospholipase B-like 1 chain C; PLB homolog 1; putative phospholipase B-like 1
Gene Aliases: 1100001H23Rik; im:7138859; PLBD1; RGD1308734; zgc:194816
UniProt ID: (Bovine) Q9GL30, (Human) Q6P4A8, (Mouse) Q8VCI0, (Rat) Q5U2V4
Entrez Gene ID: (Bovine) 317710, (Dog) 486673, (Pig) 100155493, (Horse) 100066968, (Human) 79887, (Rabbit) 100347232, (Guinea pig) 100713563, (Mouse) 66857, (Rat) 297694, (Zebrafish) 569736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.