OriGene
Manufacturer Code:TA805345
Catalog # 50-168-0659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 107-379 of human PLAU produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATF BDPLT5 QPD UPA URK u-PA; plasminogen activator, urinary; U-plasminogen activator; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A
Gene Aliases: ATF; BDPLT5; PLAU; QPD; u-PA; UPA; URK
UniProt ID: (Human) P00749
Entrez Gene ID: (Human) 5328
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.