Invitrogen
Manufacturer Code:PA534457
Catalog # PIPA534457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid synthetic peptide near the amino terminus of human PLAGL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2H2-type zinc finger protein; PLAGL2 Pleiomorphic adenoma gene-like 2 Zinc finger protein PLAGL2 ZNF900; pleiomorphic adenoma gene-like 2; Pleiomorphic adenoma-like protein 2; pleomorphic adenoma gene like 2; Zinc finger protein PLAGL2
Gene Aliases: AU018672; AW552839; KIAA0198; mKIAA0198; PLAGL2; ZNF900
UniProt ID: (Human) Q9UPG8
Entrez Gene ID: (Human) 5326, (Mouse) 54711, (Rat) 296281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.