Novus Biologicals
Manufacturer Code:NBP29324120UL
Catalog # NB028370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 240-470 of human PLAGL2 (NP_002648.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2H2-type zinc finger protein; C2H2-type zinc finger protein FLJ23283 KIAA0198 pleiomorphic adenoma gene-like 2 Pleiomorphic adenoma-like protein 2 zinc finger protein PLAGL2 ZNF900; pleiomorphic adenoma gene-like 2; Pleiomorphic adenoma-like protein 2; Zinc finger protein PLAGL2
Gene Aliases: KIAA0198; PLAGL2; ZNF900
UniProt ID: (Human) Q9UPG8
Entrez Gene ID: (Human) 5326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.