Proteintech
Manufacturer Code:122841AP150UL
Catalog # 50-172-7859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLAC8 Fusion Protein Ag2939 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C15 onzin PLAC8 placenta 8 Protein C15; down-regulated in gastrointestinal cancer protein; Onzin; placenta-specific 8; Placenta-specific gene 8 protein; Protein C15
Gene Aliases: BM-004; C15; D5Wsu111e; DGIC; onzin; PLAC8; PNAS-144
UniProt ID: (Human) Q9NZF1, (Mouse) Q9JI48
Entrez Gene ID: (Human) 51316, (Mouse) 231507, (Rat) 360914
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.