Proteintech
Manufacturer Code:602441IG150UL
Catalog # 50-173-6342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5C10G9 |
Immunogen | PLAA Fusion Protein Ag3224 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DOA1 homolog; DOA1 PLA2P PLAA PLAP; Phospholipase A-2-activating protein; phospholipase A2-activating protein; PLA2P
Gene Aliases: 2410007N06; AI536418; AU018445; AW208417; D4Ertd618e; DOA1; PLA2P; PLAA; PLAP; Ufd3
UniProt ID: (Human) Q9Y263, (Mouse) P27612, (Rat) P54319
Entrez Gene ID: (Human) 9373, (Mouse) 18786, (Rat) 116645
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.