Novus Biologicals
Manufacturer Code:NBP250248AF647
Catalog # NBP250248AH
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 12/6/05 |
Immunogen | Human PLA2R extracellular sequence N-C3 (containing domains N-terminal Cysteine rich-FibII-CTLD1-CTLD2-CTLD3). Genbank acc no. U17033 amino acids 20-663 Recombinant protein expressed in HEK 293-EBNA-1 cells |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 180 kDa secretory phospholipase A2 receptor; 180 kDa secretory phospholipase A2 receptor CLEC13CC-type lectin domain family 13 member C phospholipase A2 receptor 1 180kDa PLA2G1R PLA2IR PLA2-RM-type receptor PLA2Rphospholipase A2 receptor 1 180kD secretory phospholipase A2 receptor; C-type lectin domain family 13 member C; M-type receptor; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; PLA2-R; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor
Gene Aliases: CLEC13C; PLA2-R; PLA2G1R; PLA2IR; PLA2R; PLA2R1
UniProt ID: (Human) Q13018
Entrez Gene ID: (Human) 22925
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.