Invitrogen
Manufacturer Code:PA552966
Catalog # PIPA552966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PLA2R1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 180 kDa secretory phospholipase A2 receptor; C-type lectin domain family 13 member C; M-type receptor; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; PLA2-R; PLA2R1 phospholipase A2 receptor 1 180kDa CLEC13C PLA2-R PLA2G1R PLA2IR PLA2R phospholipase A2 receptor 1; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor
Gene Aliases: CLEC13C; PLA2-R; PLA2G1R; PLA2IR; PLA2R; PLA2R1
UniProt ID: (Human) Q13018
Entrez Gene ID: (Human) 22925
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.