Novus Biologicals
Manufacturer Code:NBP299645JF549
Catalog # NB062262
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant PLA2G7/PAF-AH/Lp-PLA2 (Q13093-1 Met1-Asn441). |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-alkyl-2-acetylglycerophosphocholine esterase; 2-acetyl-1-alkylglycerophosphocholine esterase; 2-acetyl-1-alkylglycerophosphocholine esterase EC 3.1.1 EC 3.1.1.471-alkyl-2-acetylglycerophosphocholine esterase Group-VIIA phospholipase A2 gVIIA-PLA2 LDL-associated phospholipase A2 LDL-PLA(2) LDL-PLA2 lipoprotein-associated phospholipase A2 LP-PLA2 PAF acetylhydrolase PAFAHPAF 2-acylhydrolase phospholipase A2 group VII (platelet-activating factor acetylhydrolase plasma) platelet-activating factor acetylhydrolase; Group-VIIA phospholipase A2; gVIIA-PLA2; LDL-associated phospholipase A2; LDL-PLA(2); lipoprotein-associated phospholipase A2; PAF 2-acylhydrolase; PAF acetylhydrolase; phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma); Platelet-activating factor acetylhydrolase
Gene Aliases: LDL-PLA2; LP-PLA2; PAFAD; PAFAH; PLA2G7
UniProt ID: (Human) Q15692
Entrez Gene ID: (Human) 7941
Molecular Function: esterase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.