Invitrogen
Manufacturer Code:PA542005
Catalog # PIPA542005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PLA2G5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ca2+-dependent phospholipase A2; calcium-dependent phospholipase A2; group V phospholipase A2; Phosphatidylcholine 2-acylhydrolase 5; Phospholipase A2 group V; Phospholipase A2 group V DKFZp686C2294 GV-PLA2 MGC46205 PLA2-10 hVPLA(2) FRFB; phospholipase A2, group 5; phospholipase A2, group V; PLA2-10
Gene Aliases: FRFB; GV-PLA2; hVPLA(2); PLA2-10; PLA2G5
UniProt ID: (Human) P39877, (Rat) P51433
Entrez Gene ID: (Horse) 100058536, (Bovine) 614911, (Dog) 478207, (Human) 5322, (Guinea pig) 100728430, (Rat) 29354
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.