Abnova
Manufacturer Code:H00005322K
Catalog # 89-210-049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PLA2G5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PLA2G5 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Ca2+-dependent phospholipase A2; DKFZp686C2294 GV-PLA2 MGC46205 PLA2-10 hVPLA(2); Phosphatidylcholine 2-acylhydrolase 5; Phospholipase A2 group V; phospholipase A2, group V; PLA2-10
Gene Aliases: FRFB; GV-PLA2; hVPLA(2); PLA2-10; PLA2G5
UniProt ID: (Human) P39877
Entrez Gene ID: (Human) 5322
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.