Invitrogen
Manufacturer Code:PA563344
Catalog # PIPA563344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PLA2G4F |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cPLA2-zeta; Cytosolic phospholipase A2 zeta; Phospholipase A2 group IVF; phospholipase A2, group IVF; PLA2G4FZ cPLA2-zeta cytosolic phospholipase A2 zeta
Gene Aliases: PLA2G4F; PLA2G4FZ
UniProt ID: (Human) Q68DD2
Entrez Gene ID: (Human) 255189
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.