Novus Biologicals
Manufacturer Code:NBP232610
Catalog # NBP232610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: LCLLDTAFFVNSSYPPLLRPERKADLIIHLNYCAGSQTKPLKQTCEYCTVQNIPFPKYELPDENENLKECYLMENPQEPDAPIVTFFPLIN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-dependent N-acyltransferase; cPLA2-epsilon; cPLA2-epsilon cytosolic phospholipase A2 epsilon EC 3.1.1.4 FLJ45651 MGC126633 MGC126661 Phospholipase A2 group IVE phospholipase A2 group IVE; Cytosolic phospholipase A2 epsilon; Phospholipase A2 group IVE; phospholipase A2, group IVE
Gene Aliases: PLA2G4E
UniProt ID: (Human) Q3MJ16
Entrez Gene ID: (Human) 123745
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.