Proteintech
Manufacturer Code:180881AP150UL
Catalog # 50-173-1979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLA2G4E Fusion Protein Ag12808 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-dependent N-acyltransferase; cPLA2-epsilon; Cytosolic phospholipase A2 epsilon; Phospholipase A2 group IVE; phospholipase A2 group IVE PLA2G4E PLA2G4E Ens; phospholipase A2, group 4E; phospholipase A2, group IVE
Gene Aliases: 2310026J01Rik; C230096D22; Pla2epsilon; PLA2G4E; RGD1310595
UniProt ID: (Human) Q3MJ16, (Mouse) Q50L42
Entrez Gene ID: (Human) 123745, (Rat) 296091, (Mouse) 329502
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.