Novus Biologicals
Manufacturer Code:NBP293545100UL
Catalog # NB028963
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human PLA2G4C (NP_003697.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cPLA2-gamma; cPLA2-gamma cytosolic phospholipase A2 gamma DKFZp586C0423 EC 3.1.1.4 FLJ42247 FLJ44164 Phospholipase A2 group IVC phospholipase A2 group IVC (cytosolic calcium-independent); Cytosolic lysophospholipase; Cytosolic lysophospholipid O-acyltransferase; Cytosolic phospholipase A2 gamma; Phospholipase A2 group IVC; phospholipase A2, group IVC (cytosolic, calcium-independent)
Gene Aliases: CPLA2-gamma; PLA2G4C
UniProt ID: (Human) Q9UP65
Entrez Gene ID: (Human) 8605
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.