Abnova
Manufacturer Code:PAB27669
Catalog # 89-129-182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human PLA2G4C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of PLA2G4C.
For Research Use Only
Protein Aliases: CPLA2-gamma; Cytosolic lysophospholipase; Cytosolic lysophospholipid O-acyltransferase; Cytosolic phospholipase A2 gamma; DKFZp586C0423; Phospholipase A2 group IVC; phospholipase A2, group IVC (cytosolic, calcium-independent)
Gene Aliases: CPLA2-gamma; PLA2G4C
UniProt ID: (Human) Q9UP65
Entrez Gene ID: (Human) 8605
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.