Invitrogen
Manufacturer Code:PA529100
Catalog # PIPA529100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 263 of Human PLA2G4A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium-dependent phospholipid-binding protein; cPLA2; Cytosolic phospholipase A2; Lysophospholipase; Phosphatidylcholine 2-acylhydrolase; Phospholipase A2; Phospholipase A2 group IVA; phospholipase A2, group IVA (cytosolic, calcium-dependent); PLA2G4 cPLA2-alpha cPLA2 calcium-dependent phospholipid-binding protein cytosolic phospholipase A2 lysophospholipase phosphatidylcholine 2-acylhydrolase phospholipase A2 group IVA
Gene Aliases: CPLA2; cPLA2-alpha; cPLA2alpha; PLA2G4; PLA2G4A
UniProt ID: (Human) P47712, (Mouse) P47713
Entrez Gene ID: (Human) 5321, (Mouse) 18783
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.