Invitrogen
Manufacturer Code:PA532988
Catalog # PIPA532988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 12 amino acid peptide from internal region of human PLA2G3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GIII sPLA2; GIII-SPLA2 SPLA2III GIII sPLA2 group 3 secretory phospholipase A2 group III secreted phospholipase A2 group III secretory phospholipase A2 phosphatidylcholine 2-acylhydrolase 3 phosphatidylcholine 2-acylhydrolase GIII sPLA2-III; Group 3 secretory phospholipase A2; group III secreted phospholipase A2; Group III secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 3; phosphatidylcholine 2-acylhydrolase GIII; phospholipase A2, group III
Gene Aliases: GIII-SPLA2; PLA2G3; sPLA2-III; SPLA2III
UniProt ID: (Human) Q9NZ20
Entrez Gene ID: (Human) 50487
Molecular Function: annexin calcium-binding protein calmodulin hydrolase intracellular calcium-sensing protein lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.