Novus Biologicals
Manufacturer Code:NBP133322
Catalog # NBP133322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PLA2G3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.4 GIII sPLA2 GIII-SPLA2 group 3 secretory phospholipase A2 group III secreted phospholipase A2 Group III secretory phospholipase A2 Phosphatidylcholine 2-acylhydrolase 3 phosphatidylcholine 2-acylhydrolase GIII phospholipase A2 group III sPLA2-III SPLA2III; GIII sPLA2; Group 3 secretory phospholipase A2; group III secreted phospholipase A2; Group III secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 3; phosphatidylcholine 2-acylhydrolase GIII; phospholipase A2, group III
Gene Aliases: GIII-SPLA2; PLA2G3; sPLA2-III; SPLA2III
UniProt ID: (Human) Q9NZ20
Entrez Gene ID: (Human) 50487
Molecular Function:
annexin
calcium-binding protein
calmodulin
hydrolase
intracellular calcium-sensing protein
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.