Abnova
Manufacturer Code:H00050487K
Catalog # 89-202-341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PLA2G3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PLA2G3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: GIII sPLA2; GIII-SPLA2 SPLA2III; Group 3 secretory phospholipase A2; group III secreted phospholipase A2; Group III secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 3; phosphatidylcholine 2-acylhydrolase GIII; phospholipase A2, group III
Gene Aliases: GIII-SPLA2; PLA2G3; sPLA2-III; SPLA2III
UniProt ID: (Human) Q9NZ20
Entrez Gene ID: (Human) 50487
Molecular Function: annexin calcium-binding protein calmodulin hydrolase intracellular calcium-sensing protein lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.