Novus Biologicals
Manufacturer Code:NBP273455AF532
Catalog # NP2734A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 21-144 of human PLA2G2A (NP_000291) produced in E.coli. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.1.1.4 GIIC sPLA2 Group IIA phospholipase A2 MOM1 Non-pancreatic secretory phospholipase A2 NPS-PLA2 Phosphatidylcholine 2-acylhydrolase 2A phospholipase A2 group IIA (platelets synovial fluid) phospholipase A2 membrane associated PLA2BPLA2 PLA2LPLA2S PLAS1 RASF-A sPLA2; GIIC sPLA2; Group IIA phospholipase A2; Non-pancreatic secretory phospholipase A2; NPS-PLA2; Phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2, group IIA (platelets, synovial fluid); Phospholipase A2, membrane associated
Gene Aliases: MOM1; PLA2; PLA2B; PLA2G2A; PLA2L; PLA2S; PLAS1; RASF-A; sPLA2
UniProt ID: (Human) P14555
Entrez Gene ID: (Human) 5320
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.