Invitrogen
Manufacturer Code:MA537885
Catalog # PIMA537885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2416 |
Immunogen | Recombinant protein of human PLA2G2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.4 EF Enhancing factor eted enzyme type IIA phospholipase A2 GIIC sPLA2 group IIA phospholipase A2 modifier of Min1 Mom1 non-pancreatic secreted type II phospholipase A2 non-pancreatic secretory phospholipase A2 NPS-PLA2 phosphatidylcholine 2-acylhydrolase 2A phospholipase A2 group IIA phospholipase A2 group IIA (platelets synovial fluid) phospholipase A2 membrane associated PLA2 PLA2B PLA2BPLA2 PLA2G2A PLA2L PLA2LPLA2S PLA2S PLAS1 platelet phospholipase A2 RASF-A secretory group II phospholipase A2 sPLA2 sPla2-IIA; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; non-pancreatic secreted type II phospholipase A2; Non-pancreatic secretory phospholipase A2; NPS-PLA2; Phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2, group IIA (platelets, synovial fluid); Phospholipase A2, membrane associated; platelet phospholipase A2; secretory group II phospholipase A2
Gene Aliases: EF; MOM1; PLA2; PLA2B; PLA2G2A; PLA2L; PLA2S; PLAS1; RASF-A; sPLA2; sPla2-IIA
UniProt ID: (Human) P14555, (Rat) P14423, (Mouse) P31482
Entrez Gene ID: (Human) 5320, (Rat) 29692, (Mouse) 18780
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.