Invitrogen
Manufacturer Code:PA547672
Catalog # PIPA547672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sf 21-derived recombit mouse PLA2G2A Asn22-Cys146 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: EF; Enhancing factor; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; Mom1 PLA2B PLA2L RASF-A EC 3.1.1.4 GIIC sPLA2 Group IIA phospholipase A2 MOM1 Non-pancreatic secretory phospholipase A2 NPS-PLA2 Phosphatidylcholine 2-acylhydrolase 2A phospholipase A2 group IIA (platelets synovial fluid) phospholipase A2 membrane associated PLA2BPLA2 PLA2LPLA2S PLAS1 RASF-A sPLA2; non-pancreatic secreted type II phospholipase A2; Phosphatidylcholine 2-acylhydrolase 2A; Phospholipase A2, membrane associated; secretory group II phospholipase A2
Gene Aliases: EF; Mom1; Pla2; Pla2g2a; sPLA2; sPla2-IIA
UniProt ID: (Mouse) P31482
Entrez Gene ID: (Mouse) 18780
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.