Novus Biologicals
Manufacturer Code:NBP290537R
Catalog # NB074699
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse PLA2G1B (NP_035237.1 Met 1-Cys146). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Group IB phospholipase A2; Group IB phospholipase A2 Phosphatidylcholine 2-acylhydrolase 1B phospholipase A2 phospholipase A2 group IB (pancreas) PLA2AMGC119834 PLA2EC 3.1.1.4 PPLA2MGC119835; Phosphatidylcholine 2-acylhydrolase 1B; Phospholipase A2; phospholipase A2, group IB (pancreas)
Gene Aliases: PLA2; PLA2A; PLA2G1B; PPLA2
UniProt ID: (Human) P04054
Entrez Gene ID: (Human) 5319
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.