R&D Systems
Manufacturer Code:AF6190
Catalog # AF6190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PLA2G16 Asp12-Asp132 Accession # P53816 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Adipose-specific phospholipase A2; Adipose-specific phospholipase A2 AdPLA AdPLAHRASLS3 EC 3.1.1.4 group XVI phospholipase A2 HRAS-like suppressor 3Ca-independent phospholipase A1/2 HRASLS3 HREV107 H-REV107-1 HREV107-3adipose-specific PLA2 HREV107H-rev 107 protein homolog MGC118754 phospholipase A2 group XVI Renal carcinoma antigen NY-REN-65; adipose-specific PLA2; AdPLA; Ca-independent phospholipase A1/2; Group XVI phospholipase A1/A2; group XVI phospholipase A2; H-rev 107 protein homolog; H-REV107; HRAS-like suppressor 1; HRAS-like suppressor 3; HREV107-3; HRSL3; Phospholipase A and acyltransferase 3; phospholipase A2, group XVI; Renal carcinoma antigen NY-REN-65
Gene Aliases: AdPLA; H-REV107-1; HRASLS3; HREV107; HREV107-1; HREV107-3; HRSL3; PLA2G16; PLAAT3
UniProt ID: (Human) P53816
Entrez Gene ID: (Human) 11145
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.