OriGene
Manufacturer Code:CF506912
Catalog # 50-166-4528
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C9 |
Immunogen | Full length human recombit protein of human PLA2G16 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adipose-specific phospholipase A2; adipose-specific PLA2; AdPLA; AdPLA H-REV107 H-REV107-1 HRASLS3 HREV107 HREV107-1 HREV107-3 HRSL3; Ca-independent phospholipase A1/2; Group XVI phospholipase A1/A2; group XVI phospholipase A2; H-rev 107 protein homolog; H-REV107; HRAS-like suppressor 1; HRAS-like suppressor 3; HREV107-3; HRSL3; Phospholipase A and acyltransferase 3; phospholipase A2, group XVI; Renal carcinoma antigen NY-REN-65
Gene Aliases: AdPLA; H-REV107-1; HRASLS3; HREV107; HREV107-1; HREV107-3; HRSL3; PLA2G16; PLAAT3
UniProt ID: (Human) P53816
Entrez Gene ID: (Human) 11145
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.