Invitrogen
Manufacturer Code:PA530867
Catalog # PIPA530867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 189 of Human PLA2G12A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: group XII secreted phospholipase A2; group XIIA secreted phospholipase A2; Group XIIA secretory phospholipase A2; GXII PLA2G12 ROSSY GXII sPLA2 group XII secreted phospholipase A2 group XIIA secreted phospholipase A2 group XIIA secretory phospholipase A2 phosphatidylcholine 2-acylhydrolase 12A sPLA2-XII; GXII sPLA2; Phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2, group XII; phospholipase A2, group XIIA; sPLA2-XII
Gene Aliases: FKSG38; GXII; PLA2G12; PLA2G12A; ROSSY; UNQ2519/PRO6012
UniProt ID: (Human) Q9BZM1
Entrez Gene ID: (Human) 81579
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.