Proteintech
Manufacturer Code:160091AP150UL
Catalog # 50-173-0638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLA2G12A Fusion Protein Ag8722 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: group XII secreted phospholipase A2; group XII-1 phospholipase A2; group XIIA secreted phospholipase A2; Group XIIA secretory phospholipase A2; GXII GXII sPLA2 phospholipase A2 group XIIA PLA2G12 PLA2G12A ROSSY sPLA2 XII UNQ2519/PRO6012; GXII sPLA2; Phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2, group XII; phospholipase A2, group XIIA; sPLA2-XII
Gene Aliases: 2310004B05Rik; FKSG38; GXII; GXII-1; GXII-1-PLA2; mGXII-1; PLA2G12; PLA2G12A; ROSSY; UNQ2519/PRO6012
UniProt ID: (Human) Q9BZM1, (Mouse) Q9EPR2
Entrez Gene ID: (Human) 81579, (Rat) 362039, (Mouse) 66350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.