Invitrogen
Manufacturer Code:MA529761
Catalog # PIMA529761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | Recombit Mouse PLA2G1B Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Group IB phospholipase A2; Phosphatidylcholine 2-acylhydrolase 1B; Phospholipase A2; PLA2-Ib; PLA2G1B phospholipase A2 group IB (pancreas) MGC119834 MGC119835 PLA2 PLA2A PPLA2 phospholipase A2 group IB
Gene Aliases: Pla2; Pla2a; Pla2g1b; sPLA2IB
UniProt ID: (Mouse) Q9Z0Y2
Entrez Gene ID: (Mouse) 18778
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.