Invitrogen
Manufacturer Code:PA524805
Catalog # PIPA524805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 116-145 amino acids from the C-terminal region of human PLA2G1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Group IB phospholipase A2; Phosphatidylcholine 2-acylhydrolase 1B; Phospholipase A2; phospholipase A2, group IB (pancreas); PLA2 PLA2A PPLA2 group IB phospholipase A2 phosphatidylcholine 2-acylhydrolase 1B phospholipase A2; PLA2-Ib
Gene Aliases: PLA2; PLA2A; PLA2G1B; PPLA2; sPLA2IB
UniProt ID: (Human) P04054, (Mouse) Q9Z0Y2
Entrez Gene ID: (Human) 5319, (Mouse) 18778
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.