Invitrogen
Manufacturer Code:PA518800
Catalog # PIPA518800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NKAHKNLDTKKYCQS) corresponding to the C-terminus amino acids of PLA2G1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Group IB phospholipase A2; Phosphatidylcholine 2-acylhydrolase 1B; Phospholipase A2; phospholipase A2, group IB (pancreas); PLA2G1B phospholipase A2 group IB (pancreas) MGC119834 MGC119835 PLA2 PLA2A PPLA2 phospholipase A2 group IB
Gene Aliases: PLA2; PLA2A; PLA2G1B; PPLA2
UniProt ID: (Human) P04054
Entrez Gene ID: (Human) 5319
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.