Invitrogen
Manufacturer Code:MA537667
Catalog # PIMA537667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1441CT628.33.40 |
Immunogen | PKR recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2310047A08Rik 4732414G15Rik AI467567 AI747578 double stranded RNA activated protein kinase dsRNA-activated kinase EC 2.7.11.1 eIF-2 alpha eIF2a Kinase eIF-2A protein kinase 2 EIF2AK1 EIF2AK2 eukaryotic translation initiation factor 2 alpha kinase 2 eukaryotic translation initiation factor 2-alpha kinase 2 IFN- type I-induced and dsRNA-activated kinase IFN-induced and double-stranded RNA-activated kinase interferon-induced double-stranded RNA-activated protein kinase interferon-inducible elF2alpha kinase interferon-inducible RNA-dependent protein kinase MGC126524 OTTHUMP00000201320 P1/eIF-2A protein kinase p68 kinase PKR PPP1R83 PRKR protein kinase R protein kinase RNA-activated protein kinase interferon inducible double stranded RNA dependent Protein kinase interferon-inducible double stranded RNA dependent protein phosphatase 1 regulatory subunit 83 Serine/threonine-protein kinase TIK T-cell viral integration site Tik Tyrosine-protein kinase EIF2AK2; double stranded RNA activated protein kinase; eIF-2A protein kinase 2; Eukaryotic translation initiation factor 2-alpha kinase 2; Interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; Interferon-inducible RNA-dependent protein kinase; P1/eIF-2A protein kinase; p68 kinase; PKR; protein kinase R; Protein kinase RNA-activated; protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; Tyrosine-protein kinase EIF2AK2
Gene Aliases: EIF2AK1; EIF2AK2; PKR; PPP1R83; PRKR
UniProt ID: (Human) P19525
Entrez Gene ID: (Human) 5610
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.