Invitrogen
Manufacturer Code:PA517395
Catalog # PIPA517395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the sequence of human PKR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: double stranded RNA activated protein kinase; eIF-2A protein kinase 2; Eukaryotic translation initiation factor 2-alpha kinase 2; Interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; Interferon-inducible RNA-dependent protein kinase; OTTHUMP00000201320 P1/eIF-2A protein kinase double stranded RNA activated protein kinase eIF-2A protein kinase 2 interferon-induced double-stranded RNA-activated protein kinase interferon-inducible RNA-dependent protein kinase interferon-inducible elF2alpha kinase p68 kinase protein kinase RNA-activated protein kinase interferon-inducible double stranded RNA dependent; P1/eIF-2A protein kinase; p68 kinase; PKR; protein kinase R; Protein kinase RNA-activated; protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; Tyrosine-protein kinase EIF2AK2
Gene Aliases: EIF2AK1; EIF2AK2; PKR; PPP1R83; PRKR
UniProt ID: (Human) P19525
Entrez Gene ID: (Human) 5610
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.