Novus Biologicals
Manufacturer Code:NBP176730
Catalog # NBP176730
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human PKR. The immunogen is located within the first 50 amino acids of PKR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: double stranded RNA activated protein kinase; double stranded RNA activated protein kinase EC 2.7.11.1 eIF-2A protein kinase 2 EIF2AK1 eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase interferon-induced double-stranded RNA-activated protein kinase interferon-inducible double stranded RNA dependent interferon-inducible elF2alpha kinase Interferon-inducible RNA-dependent protein kinase PKRp68 kinase PRKRMGC126524 Protein kinase RNA-activated; eIF-2A protein kinase 2; Eukaryotic translation initiation factor 2-alpha kinase 2; Interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; Interferon-inducible RNA-dependent protein kinase; P1/eIF-2A protein kinase; p68 kinase; PKR; protein kinase R; Protein kinase RNA-activated; protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; Tyrosine-protein kinase EIF2AK2
Gene Aliases: EIF2AK1; EIF2AK2; PKR; PPP1R83; PRKR
UniProt ID: (Human) P19525
Entrez Gene ID: (Human) 5610
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.