Invitrogen
Manufacturer Code:MA515887
Catalog # PIMA515887
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D1 |
Immunogen | Purified recombit fragment of human PRK2 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cardiolipin-activated protein kinase Pak2; p140 kinase; PKN gamma; protease-activated kinase 2; Protein kinase C-like 2; Protein-kinase C-related kinase 2; RP5-905H16.1 PAK2 PRK2 PRKCL2 PRO2042 Pak-2 cardiolipin-activated protein kinase Pak2 protein kinase C-like 2 protein-kinase C-related kinase 2 serine/threonine-protein kinase N2; serine/threonine kinase 7; Serine/threonine-protein kinase N2
Gene Aliases: 6030436C20Rik; AI507382; Pak-2; PAK2; PKN2; PRK2; PRKCL2; PRO2042; STK7
UniProt ID: (Human) Q9H1W4, (Mouse) Q8BWW9
Entrez Gene ID: (Human) 5586, (Mouse) 109333, (Rat) 207122
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.